Learn More
Medchemexpress LLC HY-10404 5mg Medchemexpress, Dilmapimod CAS:444606-18-2 Purity:>98%
SDP
Catalog No. 501932255
Encompass_Preferred
Medchemexpress, HY-10404 5mg Dilmapimod CAS:444606-18-2 Dilmapimod (SB-681323) is a potent p38 MAPK inhibitor that potentially suppresses inflammation in chronic obstructive pulmonary disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2255 Supplier Medchemexpress LLC Supplier No. HY104045MG
Medchemexpress, HY-10404 5mg Dilmapimod CAS:444606-18-2 Dilmapimod (SB-681323) is a potent p38 MAPK inhibitor that potentially suppresses inflammation in chronic obstructive pulmonary disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.