Learn More
Medchemexpress LLC Diloxanide | 579-38-4 | 99.7% | 100 MG
SDP
Catalog No. 5000233217
Encompass_Preferred
Diloxanide (CAS 579-38-4) is an anti-protozoal research compound supplied as a solid or DMSO solution for in vitro and analytical applications. Reported purity is approximately 99.7%, and typical formats include multiple solid pack sizes and a 10 mM solution in DMSO. Intended for research use only.
  • Anti-protozoal agent for research on intestinal amebiasis.
  • High reported purity, suitable for analytical applications.
  • Available as solid quantities and as a DMSO solution for assay use.
  • Soluble in DMSO.
  • Intended for in vitro and analytical research (research use only).
Catalog No. 50-002-33217 Supplier Medchemexpress LLC Supplier No. HY119972100MG
Diloxanide (CAS 579-38-4) is an anti-protozoal research compound supplied as a solid or DMSO solution for in vitro and analytical applications. Reported purity is approximately 99.7%, and typical formats include multiple solid pack sizes and a 10 mM solution in DMSO. Intended for research use only.
  • Anti-protozoal agent for research on intestinal amebiasis.
  • High reported purity, suitable for analytical applications.
  • Available as solid quantities and as a DMSO solution for assay use.
  • Soluble in DMSO.
  • Intended for in vitro and analytical research (research use only).