Learn More
Medchemexpress LLC Diloxanide furoate | 3736-81-0 | 99.6% | 25 MG
SDP
Catalog No. 5000414517
Encompass_Preferred
Diloxanide furoate is the proagent of Diloxanide. It is a potent and orally active anti-protozoal agent that can be used for the research of amebiasis, mild intestinal amebiasis or asymptomatic cyst carriers.
  • Potent and orally active anti-protozoal agent.
  • Can be used for the research of amebiasis, mild intestinal amebiasis or asymptomatic cyst carriers.
  • Potent activity against different strains of *E. histolytica* with MIC values ranging from 1.95 μg/ml to 2.5 μg/ml.
  • Effective against intestinal (caecal) amoebiasis in rats, with an ED50 value of 77.9 mg/kg.
Catalog No. 50-004-14517 Supplier Medchemexpress LLC Supplier No. 25MGHYB1147
Diloxanide furoate is the proagent of Diloxanide. It is a potent and orally active anti-protozoal agent that can be used for the research of amebiasis, mild intestinal amebiasis or asymptomatic cyst carriers.
  • Potent and orally active anti-protozoal agent.
  • Can be used for the research of amebiasis, mild intestinal amebiasis or asymptomatic cyst carriers.
  • Potent activity against different strains of *E. histolytica* with MIC values ranging from 1.95 μg/ml to 2.5 μg/ml.
  • Effective against intestinal (caecal) amoebiasis in rats, with an ED50 value of 77.9 mg/kg.