Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Diloxanide (furoate), 97%

Catalog No. AAH36096MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH36096MD 250 mg
AAH3609603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH36096MD Supplier Thermo Scientific™ Supplier No. H36096MD
Add to Cart

Chemical Identifiers

CAS 3736-81-0
Molecular Formula C14H11Cl2NO4
Molecular Weight (g/mol) 328.15
MDL Number MFCD00083302
InChI Key BDYYDXJSHYEDGB-UHFFFAOYSA-N
IUPAC Name 4-(2,2-dichloro-N-methylacetamido)phenyl furan-2-carboxylate
SMILES CN(C(=O)C(Cl)Cl)C1=CC=C(OC(=O)C2=CC=CO2)C=C1

Specifications

Quantity 250 mg
Formula Weight 328.15
Percent Purity 97%
Physical Form Solid
EINECSNumber 223-108-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.