Learn More
Medchemexpress LLC Diltiazem | 42399-41-7 | 99.7% | 100 MG
SDP
Catalog No. 5000405915
Encompass_Preferred
Diltiazem is an orally active L-type Ca2+ channel blocker. It demonstrates antihypertensive and antiarrhythmic effects. This compound is suitable for research concerning cardiac arrhythmia, hypertension, and angina pectoris.
  • Orally active L-type Ca2+ channel blocker.
  • Exhibits antihypertensive effects.
  • Exhibits antiarrhythmic effects.
  • Suitable for research of cardiac arrhythmia.
  • Suitable for research of hypertension.
  • Suitable for research of angina pectoris.
Catalog No. 50-004-05915 Supplier Medchemexpress LLC Supplier No. HYB0632100MG
Diltiazem is an orally active L-type Ca2+ channel blocker. It demonstrates antihypertensive and antiarrhythmic effects. This compound is suitable for research concerning cardiac arrhythmia, hypertension, and angina pectoris.
  • Orally active L-type Ca2+ channel blocker.
  • Exhibits antihypertensive effects.
  • Exhibits antiarrhythmic effects.
  • Suitable for research of cardiac arrhythmia.
  • Suitable for research of hypertension.
  • Suitable for research of angina pectoris.