Learn More
Medchemexpress LLC Diltiazem-d3 hydrochloride | 1217623-80-7 | 98.7% | 454.00 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY14656S1MG
Encompass_Preferred
Diltiazem-d3 hydrochloride is the deuterium-labeled form of Diltiazem hydrochloride, a potent Ca2+ influx inhibitor. This compound acts as a slow channel blocker or calcium antagonist, making it valuable for various research applications.
  • Deuterium-labeled compound
  • Ca2+ influx inhibitor
  • Slow channel blocker
  • Calcium antagonist
Catalog No. 50-003-60921