Learn More
Cayman Chemical Diltiazem-d3 (hydrochloride), Purity: ≥99% deuterated forms (d1-d3), CAS: 1217860-13-3, 10 mg
SDP
Catalog No. 502941994
Encompass_Preferred
An internal standard for the quantification of diltiazem by GC- or LC-MS
Catalog No. 50-294-1994 Supplier Cayman Chemical Supplier No. 3998110 MG
An internal standard for the quantification of diltiazem by GC- or LC-MS