Learn More
Selleck Chemical LLC Diltiazem HCl 10mg 33286-22-5 RG 83606 HCl
SDP
Catalog No. 506011
Encompass_Preferred
Diltiazem (Tiazac, RG 83606) HCl is a benzothiazepine derivative with vasodilating action due to its antagonism of the actions of the calcium ion in membrane functions. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-601-1 Supplier Selleck Chemical LLC Supplier No. S186510MG
Diltiazem (Tiazac, RG 83606) HCl is a benzothiazepine derivative with vasodilating action due to its antagonism of the actions of the calcium ion in membrane functions. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.