Learn More
Medchemexpress LLC HY-14656 5g , Diltiazem (hydrochloride) CRD-401 CAS:33286-22-5 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY146565G
Encompass_Preferred
Medchemexpress, HY-14656 5g Diltiazem (hydrochloride) CRD-401 CAS:33286-22-5 Formula:C22H27ClN2O4S Purity:98% Diltiazem hydrochloride is a Ca 2+ influx inhibitor (slow channel blocker or calcium antagonist). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2267