Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diltiazem Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602218 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-218 25 g Amber Glass Bottle
18-602-219 100 g Amber Glass Bottle
18-602-221 500 g Amber Glass Bottle
18-602-220 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-602-218 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D116325GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

D1163, 33286-22-5, C22H26N2O4S·HCl

Spectrum™ Chemical Diltiazem Hydrochloride, USP is a calcium channel blocker used to prevent chest pain (angina). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 33286-22-5
Molecular Formula C22H27ClN2O4S
Molecular Weight (g/mol) 450.98
InChI Key HDRXZJPWHTXQRI-BHDTVMLSSA-N
IUPAC Name hydrogen (2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl acetate chloride
SMILES [H+].[Cl-].COC1=CC=C(C=C1)[C@@H]1SC2=CC=CC=C2N(CCN(C)C)C(=O)[C@@H]1OC(C)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation +110° to +116°
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Diltiazem Hydrochloride
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.