Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diludine (Et-HE) | 1149-23-1 | MFCD00005951 | 99.7% | 253.30 g/mol | C13H19NO4 | 500 G
SDP

Catalog No. 5000300775
Encompass_Preferred

Diludine (Et-HE) is a Hantzsch ester small molecule used as an antioxidant and hydride donor in research applications. It is supplied as an off-white to yellow solid with a defined molecular formula and high purity, and should be stored protected from light for stability.

  • Reported purity of 99.7%.
  • Molecular weight 253.30 g/mol.
  • Molecular formula C13H19NO4.
  • Off-white to yellow solid appearance.
  • Useful as an antioxidant and hydride transfer reagent in research.
  • Recommended storage: protect from light; stable under recommended conditions.

Catalog No. 50-003-00775 Supplier Medchemexpress LLC Supplier No. HYW017960500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diludine (Et-HE) is a Hantzsch ester small molecule used as an antioxidant and hydride donor in research applications. It is supplied as an off-white to yellow solid with a defined molecular formula and high purity, and should be stored protected from light for stability.

  • Reported purity of 99.7%.
  • Molecular weight 253.30 g/mol.
  • Molecular formula C13H19NO4.
  • Off-white to yellow solid appearance.
  • Useful as an antioxidant and hydride transfer reagent in research.
  • Recommended storage: protect from light; stable under recommended conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.