Learn More
Meso Scale Discovery Diluent 101, 50 mL
SDP
Catalog No. NC1229542
Encompass
Diluent for detection antibody and/or calibrator and samples; contains protein, blockers, and preservatives.
Catalog No. NC1229542 Supplier Meso Scale Discovery Supplier No. R51AD3
Diluent for detection antibody and/or calibrator and samples; contains protein, blockers, and preservatives.