Learn More
Meso Scale Discovery Diluent 11 | 50 mL
SDP
Catalog No. NC1646913
Encompass
Diluent for detection antibody and/or calibrator and samples; contains protein, blockers, and preservatives.
Catalog No. NC1646913 Supplier Meso Scale Discovery Supplier No. R55BA3
Diluent for detection antibody and/or calibrator and samples; contains protein, blockers, and preservatives.