Learn More
Teknova Diluent Solution. 1000mL, Sterile.
SDP
Supplier:  Teknova D5120
Encompass
1.00% Bovine Serum Albumin(BSA), 0.05% (v/v) Tween-20, 135mM Sodium Chloride,2.7mM Potassium Chloride,4.3mM Disodium Phosphate,1.4mM Monopotassium Phosphate
Catalog No. 50-841-654