Learn More
Meso Scale Discovery Diluent 2, 200 mL, for detection antibody, calibrator, and sample dilution
SDP
Catalog No. NC2046408
Encompass
MESO SCALE DISCOVERY (MSD) Diluent 2 (R51BB-2) is a specialized reagent designed for the precise dilution of detection antibodies, calibrators, and samples in scientific research applications. This diluent is formulated with proteins, blockers, and preservatives to enhance assay performance and minimize non-specific binding while maintaining sample integrity. Intended exclusively for research use, this diluent is not suitable for diagnostics. Currently, the product is available in a made-to-order format with specific minimum order requirements.
  • Volume: 200 mL
  • Key components include proteins, blockers, and preservatives
  • Primarily used for detection antibody, calibrator, and sample dilution
  • Stored at temperatures of ≤-10 °C
  • Intended for research use only
  • Not suitable for diagnostic procedures
Catalog No. NC2046408 Supplier Meso Scale Discovery Supplier No. R51BB2
MESO SCALE DISCOVERY (MSD) Diluent 2 (R51BB-2) is a specialized reagent designed for the precise dilution of detection antibodies, calibrators, and samples in scientific research applications. This diluent is formulated with proteins, blockers, and preservatives to enhance assay performance and minimize non-specific binding while maintaining sample integrity. Intended exclusively for research use, this diluent is not suitable for diagnostics. Currently, the product is available in a made-to-order format with specific minimum order requirements.
  • Volume: 200 mL
  • Key components include proteins, blockers, and preservatives
  • Primarily used for detection antibody, calibrator, and sample dilution
  • Stored at temperatures of ≤-10 °C
  • Intended for research use only
  • Not suitable for diagnostic procedures