Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meso Scale Discovery Diluent 45, 40 mL
SDP

Catalog No. NC1581364
Encompass

Diluent 45 is a specialized reagent designed for scientific research applications, particularly in immunoassays. It is formulated to prepare detection antibodies, calibrators, and samples, ensuring consistent and reproducible results. The diluent's unique composition includes proteins, blockers, and preservatives, which enhance sample integrity and optimize assay performance.

  • Volume: 40 mL
  • Product type: Diluent
  • Primary application: Detection antibody, calibrator, and sample dilution
  • Composition: Contains protein, blockers, and preservatives
  • Storage condition: ≤-10 °C
  • Usage statement: For research use only
  • Diagnostic use: Not for diagnostic procedures
  • Supplier: MESO SCALE DISCOVERY

Catalog No. NC1581364 Supplier Meso Scale Discovery Supplier No. R50AI4
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diluent 45 is a specialized reagent designed for scientific research applications, particularly in immunoassays. It is formulated to prepare detection antibodies, calibrators, and samples, ensuring consistent and reproducible results. The diluent's unique composition includes proteins, blockers, and preservatives, which enhance sample integrity and optimize assay performance.

  • Volume: 40 mL
  • Product type: Diluent
  • Primary application: Detection antibody, calibrator, and sample dilution
  • Composition: Contains protein, blockers, and preservatives
  • Storage condition: ≤-10 °C
  • Usage statement: For research use only
  • Diagnostic use: Not for diagnostic procedures
  • Supplier: MESO SCALE DISCOVERY

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.