Learn More
Meso Scale Discovery Diluent 58, 50 mL, for Research Use Only, not for diagnostic procedures, Storage Temperature ≤-10 °C
SDP
Catalog No. NC2163566
Encompass
MESO SCALE DISCOVERY Diluent 58 is a specialized reagent formulated for research applications, supplied in a 50 mL volume. This diluent is strictly for research use and is not intended for diagnostic procedures. Optimal storage requires temperatures of ≤-10 °C. The product has transitioned to a made-to-order status, with specific minimum order quantities potentially linked to assay plate usage.
  • Supplied in a 50 mL volume
  • For research use only
  • Not for use in diagnostic procedures
  • Requires storage at temperatures of ≤-10 °C
  • Made-to-order status
  • Minimum order quantity of 25 plates for a 5-plate pack size
This item is not returnable. View return policy
Catalog No. NC2163566 Supplier Meso Scale Discovery Supplier No. R50CA2
MESO SCALE DISCOVERY Diluent 58 is a specialized reagent formulated for research applications, supplied in a 50 mL volume. This diluent is strictly for research use and is not intended for diagnostic procedures. Optimal storage requires temperatures of ≤-10 °C. The product has transitioned to a made-to-order status, with specific minimum order quantities potentially linked to assay plate usage.
  • Supplied in a 50 mL volume
  • For research use only
  • Not for use in diagnostic procedures
  • Requires storage at temperatures of ≤-10 °C
  • Made-to-order status
  • Minimum order quantity of 25 plates for a 5-plate pack size