Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arkray Diluent 80
SDP

Catalog No. 21377292
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-377-292 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Handle with care. Avoid contact with skin or eyes. Use in a well-ventilated area.
Catalog No. 21-377-292 Supplier Arkray Supplier No. 103399
Add to Cart
Add to Cart
Handle with care. Avoid contact with skin or eyes. Use in a well-ventilated area.

Arkray Diluent 80 ensures consistent sample preparation and accurate results. Optimized for Arkray analyzers, it supports reliable performance, simplifies workflows, and maintains reproducibility in routine laboratory testing.

  • Consistent Performance: Arkray Diluent 80 provides reliable and reproducible results, ensuring confidence in every analysis.
  • Optimized for Arkray Systems: Specifically formulated to work seamlessly with Arkray analyzers, maximizing accuracy and efficiency.
  • Ready-to-Use Solution: Pre-prepared diluent saves time and reduces preparation errors, enabling immediate application in routine workflows.
  • Maintains Sample Integrity: Ensures accurate sample dilution without affecting analyte stability or measurement quality.
  • Workflow Efficiency: Simplifies laboratory operations with minimal preparation, reducing downtime and increasing productivity.
  • Wide Application: Suitable for a variety of Arkray assays, providing versatility for multiple testing needs.
  • Batch Consistency: Manufactured under strict quality controls, ensuring uniform performance across different batches.
  • Stable and Dependable: Maintains chemical stability under standard laboratory storage conditions.
  • Minimized Error: Supports reproducible results across different operators and instruments.
  • Trusted Brand: Backed by Arkray's legacy of quality and reliability in clinical diagnostics.

Specifications

Type Solution
Form Liquid

Arkray Diluent 80 ensures consistent sample dilution for accurate and reliable results. Optimized for Arkray analyzers, it streamlines workflow and supports reproducible laboratory testing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.