Learn More
MOLECULAR DEPOT LLC Diluent A - 2 5 mL
Catalog No. 5000013341
Encompass_Preferred
Diluent A is the special diluent used in Interference Test Kit (K2010001) and Interference Test Kit EXTRA (K2010008) This diluent can be spiked into the control specimen when testing the interference of Conjugated Bilirubin Ascorbic Acid Hemoglobin Serum Proteins Triglycerides Creatinine Glucose Glycerol EDTA Intralipids Urea and any other interference substance provided by Molecular Depot except Free Bilirubin and Uric Acid For in vitro research use only
Catalog No. 50-000-13341 Supplier MOLECULAR DEPOT LLC Supplier No. K2010001G
Diluent A is the special diluent used in Interference Test Kit (K2010001) and Interference Test Kit EXTRA (K2010008) This diluent can be spiked into the control specimen when testing the interference of Conjugated Bilirubin Ascorbic Acid Hemoglobin Serum Proteins Triglycerides Creatinine Glucose Glycerol EDTA Intralipids Urea and any other interference substance provided by Molecular Depot except Free Bilirubin and Uric Acid For in vitro research use only