Learn More
MOLECULAR DEPOT LLC Diluent B - 25 mL
Catalog No. 5000013442
Encompass_Preferred
Diluent B for Interference Testing (25 mL) is the special diluent used in Interference Test Kit (K2010001) This diluent should be spiked into the control specimen when testing the interference of Free Bilirubin It can also be used to dilute the stock solution of free bilirubin For in vitro research use only
Catalog No. 50-000-13442 Supplier MOLECULAR DEPOT LLC Supplier No. K2010001H
Diluent B for Interference Testing (25 mL) is the special diluent used in Interference Test Kit (K2010001) This diluent should be spiked into the control specimen when testing the interference of Free Bilirubin It can also be used to dilute the stock solution of free bilirubin For in vitro research use only