Learn More
Interscience DiluFlow BagOpen 400
SDP
Catalog No. 16948947 Shop All Interscience Products
Change view
Click to view available options
For Use With (Equipment):
400 mL Bags
Catalog No. For Use With (Equipment)
16-948-947 400 mL Bags
1 options
Catalog No. 16-948-947 Supplier Interscience Supplier No. 505016
Add to Cart
Add to Cart
The BagOpen makes it easy to open bags and keep them open in a vertical position, without any risk of contamination during handling.
  • Stand for 400 mL bags
  • Stainless steel
Specifications
Product Type Bag Opener Kit
Autoclavable Not Autoclavable
For Use With (Equipment) 400 mL Bags
Sterility Non-sterile