Learn More
Interscience DiluFlow Pro Double Pump
SDP
Catalog No. 16949018 Shop All Interscience Products
Change view
Click to view available options
Voltage:
100/240 V
Catalog No. Voltage
16-949-018 100/240 V
1 options
Catalog No. 16-949-018 Supplier Interscience Supplier No. 502205
Add to Cart
Add to Cart
Request a Quote
The DiluFlow Pro double pump is a gravimetric dilutor that automatically weighs a solid sample up to 5 kg in seconds.
The DiluFlow Pro double pump automatically dilutes a solid sample in seconds, with the appropriate weight of diluent, up to a total weight of 5 kg.
Specifications
Product Type Gravimetric Dilutor
Autoclavable Not Autoclavable
Voltage 100/240 V