Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interscience DiluFlow Tubings with Straight Connectors
SDP

Catalog No. 16948886 Shop All Interscience Products
Change view
Click to view available options
Diameter (Metric) Inner:
3.2 mm
6.4 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
16-948-886 6.4 mm
16-948-893 3.2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-948-886 Supplier Interscience Supplier No. 513044
Add to Cart
Add to Cart

Set of 5 tubings for DiluFlow

  • Set of 5 dispensing tubings (silicone and Tygon™)
  • Straight connectors
  • Inner dia. of 3.2 or 6.4 mm

Specifications

Autoclavable Autoclavable
Diameter (Metric) Inner 6.4 mm
For Use With DiluFlow
Sterility Non-sterile
Product Type Tubing
Quantity 5/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.