Learn More
Thermo Scientific™ Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Catalog No. DB4002A
Change view
Click to view available options
Product Type:
Calibration Weight
Foot Pedal
Nozzles
Printer Roll
Printer Stickers
Trocar
Weight
Packaging Quantity:
1 Ea.
2 Rolls
2 Rolls, 980/Roll
5 pieces/Bag
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Packaging Quantity
DB4002A Trocar 1 Ea.
DB4006A Calibration Weight 1 Ea.
DB4007A Foot Pedal 1 Ea.
DB4009A Nozzles 5 pieces/Bag
NC3354991 Weight 1 Ea.
DB4016A Printer Stickers 2 Rolls, 980/Roll
DB4017A Printer Roll 2 Rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. DB4002A Supplier Thermo Scientific™ Supplier No. DB4002A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Accurately dispense media and achieve powerful, quiet, reliable homogenization with maximum productivity in your sample preparation.
Achieve maximum productivity with the Thermo Scientific™ Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories. Expand the flexibility and maintain long-term functionality of the two platforms with accessories that enable the use of different media formats (bottles/bags), enhance the user interface and support proper function of the system.
The easy-to-use Diluflux Gravimetric Dilutor allows the simultaneous use of up to two diluents by having dual dispensing heads that are fed by dual peristaltic pumps mounted on the unit. The reliable and powerful Homogenizer Laboratory Blender enables rapid and efficient sample homogenization with a small footprint and quiet operation. The accessories listed here enable:
  • Alternative receptacles (bottles instead of bags)
  • Alternative media sources (broth bags/bottles)
  • Additional/replacement bag stands for ease of preparation
  • Bag storage for incubation
  • Sample labeling
  • Maintenance with replacement parts
Contact us to arrange a demonstration of the Dilutor, the Blender, and/or its accessories.
Not all products are available for sale in all territories. Please inquire.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Trocar
For Use With (Equipment) Diluflux™ Diluter and Homogenizer
Packaging Quantity 1 Ea.
For research use only.