Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DiluFlux™ Pro Automated Gravimetric Dilutor

Catalog No. DB4111A
Change view
Click to view available options
Includes:
Robotic Dispensing Arm
Simple Dispensing Arm
Model:
DiluFlux Pro-3
DiluFlux Pro-5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Model
DB4111A Simple Dispensing Arm DiluFlux Pro-3
DB4150A Robotic Dispensing Arm DiluFlux Pro-5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DB4111A Supplier Thermo Scientific™ Supplier No. DB4100A/DUP
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

Accurately dispense media into suitable containers according to a set dilution using this gravimetric diluter which allows the use of two diluents.

Accurately dispense broth medium into suitable containers for food testing using the Thermo Scientific™ DiluFlux™ Pro Automated Gravimetric Dilutor. DiluFlux Pro Dilutors are compact, easy-to-use, and designed to deliver precise quantities of diluents rapidly with the touch of a button, so that you can work more confidently and accurately. When combined with the Thermo Scientific™ Peristaltic Pump and Thermo Scientific™ Homogenizer, your lab bench will be equipped with a flexible automated culture media solution to increase your productivity.

Automate the dilution step of your Thermo Scientific™ Dry-Bags™ Enrichment Media, as well as other formats, for maximum efficiency. The easy-to-use DiluFlux Pro Gravimetric Dilutors allow the simultaneous use of two diluents by having dual dispensing heads that are fed by dual peristaltic pumps mounted into the unit.

  • Intuitive: simply press two keys, tare and go, and the DiluFlux Pro Dilutor will automatically dilute your sample to the desired dilution factor
  • Accurate: featuring Watson-Marlow™ peristaltic pumps, high-tech flow regulators, and top of the line weighing platforms for trusted reproducibility and repeatability
  • Versatile: compatible with many prepared media formats, the easy-to clean, stainless steel DiluFlux Pro Dilutor is an ideal fit for any food testing laboratory
  • High-quality: the DiluFlux Pro Dilutors are ISO 7218, ISO 6887-1, FDA BAM-compliant and are made in France under ISO 9001:2008 certification

Not all products are available for sale in all territories. Please inquire.

TRUSTED_SUSTAINABILITY

Specifications

Description DiluFlux Pro-3 Automated Gravimetric Dilutor w/ simple (non-robotic) dispensing arm
For Use With Dry-Bags
Includes Simple Dispensing Arm
Material Stainless Steel (Case)
Model DiluFlux Pro-3
Dimensions (L x W x H) 41.5 x 31 x 40 cm
Overall Length (Metric) 41.5 cm
Overall Width (Metric) 31 cm
Overall Height (Metric) 40 cm
Shipping Weight (Metric) 10.0 kg
Electrical Requirements 100/127 V 220/240 V 50/60 Hz
Voltage 100/240 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.