Learn More
Dilux LLC DILUS DILUTION RESERVOIR 20/PK
Catalog No. NC9034365
Encompass_Preferred
DiluxDilution Reservoir w/lid, sterile, individually wrapped, polypropylene, autoclavable, specially designed to dispense separate dilutions simultaneously, each of the 12 numbered chambers holds 5 ml, semi-circular chambers permit complete sample withdrawal.
Catalog No. NC9034365 Supplier Dilux LLC Supplier No. D100220PK
DiluxDilution Reservoir w/lid, sterile, individually wrapped, polypropylene, autoclavable, specially designed to dispense separate dilutions simultaneously, each of the 12 numbered chambers holds 5 ml, semi-circular chambers permit complete sample withdrawal.