Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dilute Mixed Acids Hydrochloric 5%, Nitric 5% (Trace Metal Grade), Reagents

Catalog No. 01335433
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-433 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-433 Supplier Reagents Holdings Llc Supplier No. CD00290020F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dilute Mixed Acids Hydrochloric 5%, Nitric 5% (Trace Metal Grade), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Colorless to Light Yellow
Boiling Point 100°C
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.03
pH <1
Quantity 20 L
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Dilute Mixed Acids Hydrochloric 5%, Nitric 5% (Trace Metal Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.