Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diluted Hydrochloric Acid, 10% (w/v), NF, 9.5-150%, Spectrum™ Chemical
SDP

Catalog No. p-7157589 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
20 L
4 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Poly Drum
Poly-Coated Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-224 500 mL Amber Glass Bottle
18-602-222 2.5 L Poly-Coated Glass Bottle
18-602-225 6 x 500 mL Amber Glass Bottle
18-602-223 4 x 2.5 L Poly-Coated Glass Bottle
18-602-226 20 L Poly Drum
18-602-227 200 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-602-224 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HY105500MLGL
Only null left
Add to Cart
Add to Cart

HY105, 7647-01-0, HCl

Spectrum™ Chemical Diluted Hydrochloric Acid, 10 Percent (w/v), NF is a pH acidifier excipient in pharmaceutical preparations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

CAS Max % 10%, 90%
Packaging Amber Glass Bottle
Quantity 500 mL
UN Number UN1789
Percent Purity 9.5 to 10.5%
Grade NF
Chemical Name or Material Hydrochloric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.