Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diluting Fluid for CSF (Cerebrospinal Fluid), LabChem™

Catalog No. LC135857 Shop All LabChem Products
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC135857 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC135857 Supplier LabChem, Inc. Supplier No. LC135857
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Diluting Fluid for CSF (Cerebrospinal Fluid)
Color Purple
CAS Max % 89.92, 9.88, 0.2
CAS 64-19-7, 548-62-9
Linear Formula C25H30CIN3
Density 1.01g/mL
Solubility Information Soluble in water
Formula Weight 407.99
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.01g/mL
Viscosity 1.2cSt
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Quantity 125 mL
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.