Learn More
Sigma Aldrich Fine Chemicals Biosciences Dilution bottle Butterfiel
Catalog No. 502901408
Encompass
Whatman sterile dilution bottles are designed for sample dilution of water dairy products foods and pharmaceuticals prior to microbiological testing. final pH for all SOLUTIONons is 7.2 pH /- 0.2 pH at 25C. The buffers are supplied in an easy open flip-top plastic container with a tamper-evident seal.
Catalog No. 50-290-1408 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z69946272EA
Whatman sterile dilution bottles are designed for sample dilution of water dairy products foods and pharmaceuticals prior to microbiological testing. final pH for all SOLUTIONons is 7.2 pH /- 0.2 pH at 25C. The buffers are supplied in an easy open flip-top plastic container with a tamper-evident seal.