Learn More
Description
Description
- contains monobasic potassium phosphate and is used extensively in the food, dairy, and pharmaceutical industries. Offered in 90 ml and 99 ml volumes for easy 1:10 and 1:100 dilutions. Recommended as a general diluent in laboratory procedures by the Federal Drug Administrations and in the Bacteriological Analytical Manual. This product is prepared according to Standard Methods for the Examination of Water and Wastewater for use in water testing.
- Prefilled sterile dilution bottles are designed for sample dilution of water, dairy products, foods, and pharmaceuticals prior to microbiological testing. Final pH for all solutions is 7.2 pH ±0.2 pH at 25°C.
- Easy open, flip-top, plastic container with a tamper-evident seal.
Features and Benefits
- Pre-filled and Sterile.
- Tamper evident sterility strip is integrated as part of the cap. It is much easier to open and use. Never hassle with shrink bands again
- Room temperature Storage
- Long shelf life
- Chemically inert
- Resistant to sample
Applications
- For sample dilution of water, dairy products, foods and pharmaceuticals prior to microbiological testing.
- Used extensively in the food, dairy and pharmaceutical industries.
See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.
Specifications
Specifications
| Chemical Name or Material | Butterfield's Buffer |
| pH | 7.2 |
| Quantity | 72 x 90 mL |
| Solution Type | Buffer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.