Learn More
Bio X Cell Dilution Buffer, InVivoPure, pH 8.0T, 50 mL
Catalog No. 5000666299
Encompass_Preferred
InVivoPure™ dilution buffers are specifically formulated and tested to satisfy the stringent requirements for in vivo applications. They are extremely low in endotoxin, have been screened for murine pathogens, tested in animal models for toxicity, and are formulated with respect to buffer composition and pH to satisfy the requirements of Bio X Cell's antibodies.
  • Specifically formulated and tested for in vivo applications
  • Extremely low in endotoxin (<0.5 EU/mL)
  • Screened for murine pathogens (negative results)
  • Nontoxic and nonantigenic in animal models
  • Sterile (0.2 μM filtered)
  • Does not contain calcium, magnesium, phenol red, or preservatives such as azide
  • Composition: 35 mM Na2HPO4, 1.7 mM NaH2PO4, 136 mM NaCl, 0.01% TWEEN® 80
Catalog No. 50-006-66299 Supplier Bio X Cell Supplier No. IPT080R050ML
InVivoPure™ dilution buffers are specifically formulated and tested to satisfy the stringent requirements for in vivo applications. They are extremely low in endotoxin, have been screened for murine pathogens, tested in animal models for toxicity, and are formulated with respect to buffer composition and pH to satisfy the requirements of Bio X Cell's antibodies.
  • Specifically formulated and tested for in vivo applications
  • Extremely low in endotoxin (<0.5 EU/mL)
  • Screened for murine pathogens (negative results)
  • Nontoxic and nonantigenic in animal models
  • Sterile (0.2 μM filtered)
  • Does not contain calcium, magnesium, phenol red, or preservatives such as azide
  • Composition: 35 mM Na2HPO4, 1.7 mM NaH2PO4, 136 mM NaCl, 0.01% TWEEN® 80