Learn More
Grainger Diversey Dispensing and Dosing Equipment, QuattroSelect, Wall-Mount, 4-Chemical, 1 ea
Catalog No. 502778710
Encompass_Preferred
The QuattroSelect dispenser is a wall-mount system designed to accurately dilute and dispense cleaning chemicals, eliminating the need for manual measuring or mixing while ensuring tamper-proof operation. This system supports four products and provides high and low flow rates for versatile cleaning applications.
  • Features a closed-door lockable cabinet for security
  • Includes safe-gap backflow prevention compliant with ASSE 1055B
  • Equipped with Smartvalve technology to automatically select the correct flow rate
  • Supports filling of bottles, buckets, and auto scrubbers
  • Includes a bucket fill hose measuring 8 ft.
  • Features rugged steel doors with integrated product identification windows
  • Dimensions are 52.2 cm (20.56 in.) wide by 69.85 cm (27.5 in.) high by 24.77 cm (9.75 in.) deep
Catalog No. 50-277-8710 Supplier Grainger Supplier No. 19MP89
The QuattroSelect dispenser is a wall-mount system designed to accurately dilute and dispense cleaning chemicals, eliminating the need for manual measuring or mixing while ensuring tamper-proof operation. This system supports four products and provides high and low flow rates for versatile cleaning applications.
  • Features a closed-door lockable cabinet for security
  • Includes safe-gap backflow prevention compliant with ASSE 1055B
  • Equipped with Smartvalve technology to automatically select the correct flow rate
  • Supports filling of bottles, buckets, and auto scrubbers
  • Includes a bucket fill hose measuring 8 ft.
  • Features rugged steel doors with integrated product identification windows
  • Dimensions are 52.2 cm (20.56 in.) wide by 69.85 cm (27.5 in.) high by 24.77 cm (9.75 in.) deep