The 4 + 1 Select Proportioner accommodates up to five concentrated products, diluting them into various strengths for diverse cleaning tasks. It features easy product dilution, is user-friendly, and incorporates AccuPro Pressure Regulation Technology for accurate mixing regardless of water pressure fluctuations.
- Accurately dilutes concentrated cleaning products
- Eliminates dilution variance via AccuPro Pressure Regulation Technology
- Features 4 low flow (1 GPM) bottle and 1 high flow (3.5 GPM) bucket fill outputs
- Offers 1.5–3.5 GPM flow rate options
- Eductor backflow protection (ASSE 1055 approved)
- Includes 14 pre-set metering tips (1:2.6–1:447 dilution) and one customizable tip
- Bucket fill hose (4-Foot) and storage hook included
- Tool-free maintenance and quick mounting