Learn More
Andwin Scientific DILUTION VIALS 250/CS
SDP
Catalog No. NC9480336
Encompass_Preferred
The Hematology Dilution Vials give accurate hemolysis and distortion-free cell and particle counts. This product features a one-piece polystyrene design with rounded seams, a frosted writing area, and an optically clear parallel sides that allows superior light transmission. The caps with convenient tabs for quick and easy removal also has a leakproof design. This is sold in a case of 250.
Catalog No. NC9480336 Supplier Andwin Scientific Supplier No. 2015266050
The Hematology Dilution Vials give accurate hemolysis and distortion-free cell and particle counts. This product features a one-piece polystyrene design with rounded seams, a frosted writing area, and an optically clear parallel sides that allows superior light transmission. The caps with convenient tabs for quick and easy removal also has a leakproof design. This is sold in a case of 250.