Learn More
Thomas Scientific 12 Channel Reservoir, Round Bottom, 5mL/Well, with Lid, Sterile, 20/pk
SDP
Catalog No. NC9012923
Encompass_Preferred
Reservoirs are made of medical grade, non-leachable and chemically resistant polypropylene. Each of the 12 chambers holds up to 5ml and is numbered for accurate reference.
Catalog No. NC9012923 Supplier Thomas Scientific Supplier No. 1149B21
Reservoirs are made of medical grade, non-leachable and chemically resistant polypropylene. Each of the 12 chambers holds up to 5ml and is numbered for accurate reference.