Learn More
Dilux LLC DILUX DELUTION RESRVR NS 50/PK
Catalog No. NC9819110
Encompass_Preferred
DiluxDilution Reservoir w/lid, non-sterile, polypropylene, autoclavable, specially designed to dispense separate dilutions simultaneously, each of the 12 numbered chambers holds 5 ml, semi-circular chambers permit complete sample withdrawal.
Catalog No. NC9819110 Supplier Dilux LLC Supplier No. D1003
DiluxDilution Reservoir w/lid, non-sterile, polypropylene, autoclavable, specially designed to dispense separate dilutions simultaneously, each of the 12 numbered chambers holds 5 ml, semi-circular chambers permit complete sample withdrawal.