Learn More
Cole-Parmer Stainless Steel Dewar, 3 L
Supplier:  Cole-Parmer 0377409
Encompass_Preferred
These vacuum-insulted stainless steel Dewar flasks are designed for ultra-low or very high temperature applications. Constructed entirely of stainless steel, these flasks are rugged, light, and compact.
Catalog No. NC1496719