Learn More
Cole-Parmer Stainless Steel Dewar, 1 L
Supplier:  Cole-Parmer 0377405
Encompass_Preferred
Stainless Steel Dewar, 1 L, Safely store and transport low-temperature liquid gasesSpecifically designed for applications where glass cannot be usedSafe to use with liquid nitrogen and liquid oxygen
Catalog No. NC1471570