Learn More
Medchemexpress LLC DIM-C-pPhCO2Me | 151358-48-4 | 99.5% | 380.44 | 50 MG
SDP
Catalog No. 5000439369
Encompass_Preferred
DIM-C-pPhCO2Me is a nuclear receptor 4A1 (NR4A1) antagonist that induces apoptosis and decreases various proteins including PAX3-FOXO1A, N-Myc, Rassf4, MyoD1, Grem1, and DAPK1. It also reduces TXNDC5 and IDH1 expression, while inducing ER stress markers (CHOP, ATF4, and p-PERK). This compound inhibits renal cell carcinoma and breast cancer, and is useful in rhabdomyosarcoma research.
  • Induces apoptosis
  • Decreases PAX3-FOXO1A, N-Myc, Rassf4, MyoD1, Grem1, and DAPK1 proteins
  • Reduces TXNDC5 and IDH1 expression
  • Induces markers of ER stress
  • Inhibits renal cell carcinoma
  • Inhibits breast cancer
  • Useful in rhabdomyosarcoma research
Catalog No. 50-004-39369 Supplier Medchemexpress LLC Supplier No. HY11205650MG
DIM-C-pPhCO2Me is a nuclear receptor 4A1 (NR4A1) antagonist that induces apoptosis and decreases various proteins including PAX3-FOXO1A, N-Myc, Rassf4, MyoD1, Grem1, and DAPK1. It also reduces TXNDC5 and IDH1 expression, while inducing ER stress markers (CHOP, ATF4, and p-PERK). This compound inhibits renal cell carcinoma and breast cancer, and is useful in rhabdomyosarcoma research.
  • Induces apoptosis
  • Decreases PAX3-FOXO1A, N-Myc, Rassf4, MyoD1, Grem1, and DAPK1 proteins
  • Reduces TXNDC5 and IDH1 expression
  • Induces markers of ER stress
  • Inhibits renal cell carcinoma
  • Inhibits breast cancer
  • Useful in rhabdomyosarcoma research