Learn More
Medchemexpress LLC DIM-C-pPhOCH3 | 33985-68-1 | 99.4% | 352.43 | 1 ML
SDP
Catalog No. 5000345734
Encompass_Preferred
DIM-C-pPhOCH3 is a Nur77 agonist. Nerve growth factor-induced Bα (NGFI-Bα, Nur77) is an orphan nuclear receptor. This product is for research use only.
  • Decreases survival and induces apoptosis in RKO colon cancer cells, accompanied by induction of tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) protein.
  • Induces Nur77-independent apoptosis.
  • Inhibits cell growth after treatment for 24, 48, or 72 h, with maximum inhibitory response observed after 72 h, leading to considerable cell detachment and dead cells.
  • Growth-inhibitory effects after 72 h are accompanied by markers of apoptosis, including PARP cleavage and cleavage of caspase-3, caspase-9, and caspase-8.
  • Inhibits tumor growth in athymic nude mice bearing RKO cell xenografts.
  • Significantly decreases tumor volumes and final tumor weights compared with corn oil controls in athymic nude mice bearing RKO cell xenografts.
Catalog No. 50-003-45734 Supplier Medchemexpress LLC Supplier No. HY11149250MG
DIM-C-pPhOCH3 is a Nur77 agonist. Nerve growth factor-induced Bα (NGFI-Bα, Nur77) is an orphan nuclear receptor. This product is for research use only.
  • Decreases survival and induces apoptosis in RKO colon cancer cells, accompanied by induction of tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) protein.
  • Induces Nur77-independent apoptosis.
  • Inhibits cell growth after treatment for 24, 48, or 72 h, with maximum inhibitory response observed after 72 h, leading to considerable cell detachment and dead cells.
  • Growth-inhibitory effects after 72 h are accompanied by markers of apoptosis, including PARP cleavage and cleavage of caspase-3, caspase-9, and caspase-8.
  • Inhibits tumor growth in athymic nude mice bearing RKO cell xenografts.
  • Significantly decreases tumor volumes and final tumor weights compared with corn oil controls in athymic nude mice bearing RKO cell xenografts.