Learn More
Medchemexpress LLC DIM-C-pPhOCH3 | 33985-68-1 | 99.4% | 352.43 | 25 MG
SDP
Catalog No. 5000345968
Encompass_Preferred
DIM-C-pPhOCH3 is a Nur77 agonist. Nur77 (Nerve growth factor-induced Bα, NGFI-Bα) is an orphan nuclear receptor. It is used for research purposes only. In vitro, DIM-C-pPhOCH3 decreases survival and induces apoptosis in RKO colon cancer cells, accompanied by the induction of tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) protein. It also induces Nur77-independent apoptosis. In vivo, DIM-C-pPhOCH3 inhibits tumor growth in athymic nude mice bearing RKO cell xenografts.
  • Nur77 agonist
  • Induces apoptosis in RKO colon cancer cells
  • Inhibits tumor growth in athymic nude mice bearing RKO cell xenografts
  • Suitable for research applications
Catalog No. 50-003-45968 Supplier Medchemexpress LLC Supplier No. 25MGHY111492
DIM-C-pPhOCH3 is a Nur77 agonist. Nur77 (Nerve growth factor-induced Bα, NGFI-Bα) is an orphan nuclear receptor. It is used for research purposes only. In vitro, DIM-C-pPhOCH3 decreases survival and induces apoptosis in RKO colon cancer cells, accompanied by the induction of tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) protein. It also induces Nur77-independent apoptosis. In vivo, DIM-C-pPhOCH3 inhibits tumor growth in athymic nude mice bearing RKO cell xenografts.
  • Nur77 agonist
  • Induces apoptosis in RKO colon cancer cells
  • Inhibits tumor growth in athymic nude mice bearing RKO cell xenografts
  • Suitable for research applications