Learn More
Medchemexpress LLC Dim-c-pphoch3 | 33985-68-1 | 99.4% | 352.43 | 5 G
SDP
Catalog No. 5000345584
Encompass_Preferred
DIM-C-pPhOCH3 is a Nur77 agonist. Nerve growth factor-induced Bα (NGFI-Bα, Nur77) is an orphan nuclear receptor. This compound has demonstrated significant biological activity:
  • Decreases survival and induces apoptosis in RKO colon cancer cells.
  • Induces tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) protein.
  • Inhibits cell growth and induces apoptosis markers such as PARP cleavage.
  • Inhibits tumor growth in athymic nude mice bearing RKO cell xenografts.
  • Soluble in DMSO for in vitro studies.
  • Suitable for shipping at room temperature in continental US.
  • Provides long-term storage stability as powder and in solvent.
Catalog No. 50-003-45584 Supplier Medchemexpress LLC Supplier No. HY11149210MG
DIM-C-pPhOCH3 is a Nur77 agonist. Nerve growth factor-induced Bα (NGFI-Bα, Nur77) is an orphan nuclear receptor. This compound has demonstrated significant biological activity:
  • Decreases survival and induces apoptosis in RKO colon cancer cells.
  • Induces tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) protein.
  • Inhibits cell growth and induces apoptosis markers such as PARP cleavage.
  • Inhibits tumor growth in athymic nude mice bearing RKO cell xenografts.
  • Soluble in DMSO for in vitro studies.
  • Suitable for shipping at room temperature in continental US.
  • Provides long-term storage stability as powder and in solvent.