Learn More
Medchemexpress LLC DIM-C-pPhOCH3 | 33985-68-1 | 99.41% | 352.43 | 10 MG
SDP
Catalog No. 5000345709
Encompass_Preferred
DIM-C-pPhOCH3 is a Nur77 agonist. Nerve growth factor-induced Bα (NGFI-Bα, Nur77) is an orphan nuclear receptor. This product is for research use only.
  • Decreases survival and induces apoptosis in RKO colon cancer cells.
  • Induces tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) protein.
  • Inhibits cell growth after 24, 48, or 72 h.
  • Inhibits tumor growth in athymic nude mice bearing RKO cell xenografts.
Catalog No. 50-003-45709 Supplier Medchemexpress LLC Supplier No. HY11149210MM/1ML
DIM-C-pPhOCH3 is a Nur77 agonist. Nerve growth factor-induced Bα (NGFI-Bα, Nur77) is an orphan nuclear receptor. This product is for research use only.
  • Decreases survival and induces apoptosis in RKO colon cancer cells.
  • Induces tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) protein.
  • Inhibits cell growth after 24, 48, or 72 h.
  • Inhibits tumor growth in athymic nude mice bearing RKO cell xenografts.