Learn More
Medchemexpress LLC HY-111492 5mg Medchemexpress, DIM-C-pPhOCH3 CAS:33985-68-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1114925MG
Encompass_Preferred
Medchemexpress, HY-111492 5mg DIM-C-pPhOCH3 CAS:33985-68-1 DIM-C-pPhOCH3 is a Nur77 agonist. Nerve growth factor-induced Bα (NGFI-Bα, Nur77) is an orphan nuclear receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8885