Learn More
Apexbio Technology LLC DIM-C-pPhOH 151358-47-3 10mg
SDP
Catalog No. 5000096326
Encompass_Preferred
DIM-C-pPhOH (CAS 151358-47-3) is a small-molecule antagonist that targets the nuclear receptor NR4A1 (nuclear receptor subfamily 4 group A member 1) It has been shown to inhibit tumor cell proliferation by modulating the mTOR signaling pathway induce apoptosis and activate cellular stress responses In in vitro studies using human renal carcinoma cell lines ACHN and 786-O DIM-C-pPhOH demonstrated inhibitory effects on cell proliferation with IC50 values of 13 6 M and 13 0 M respectively This compound serves as a valuable tool for investigating tumor-associated signaling pathways and the mechanisms underlying anticancer therapeutics
Catalog No. 50-000-96326 Supplier Apexbio Technology LLC Supplier No. A464410
DIM-C-pPhOH (CAS 151358-47-3) is a small-molecule antagonist that targets the nuclear receptor NR4A1 (nuclear receptor subfamily 4 group A member 1) It has been shown to inhibit tumor cell proliferation by modulating the mTOR signaling pathway induce apoptosis and activate cellular stress responses In in vitro studies using human renal carcinoma cell lines ACHN and 786-O DIM-C-pPhOH demonstrated inhibitory effects on cell proliferation with IC50 values of 13 6 M and 13 0 M respectively This compound serves as a valuable tool for investigating tumor-associated signaling pathways and the mechanisms underlying anticancer therapeutics