Learn More
Medchemexpress LLC Dim-c-pphoh | 151358-47-3 | 99.0% | 338.40 | 1 ML
SDP
Catalog No. 5000439126
Encompass_Preferred
DIM-C-pPhOH is an antagonist of nuclear receptor 4A1 (NR4A1). It inhibits cancer cell growth, mTOR signaling, and induces apoptosis and cellular stress. It also reduces cell proliferation with IC50 values of 13.6 μM for ACHN cells and 13.0 μM for 786-O cells.
  • NR4A1 antagonist.
  • Inhibits cancer cell growth and mTOR signaling.
  • Induces apoptosis and cellular stress.
  • Reduces cell proliferation in ACHN and 786-O cells.
  • Inhibits NR4A1-regulated expression of survivin, bcl-2, and EGFR.
  • Activates AMPKα and inhibits mTOR.
  • Decreases expression of β1-integrin protein and mRNA.
  • Inhibits migration of MDA-MB-231 and SKBR3 cell lines.
  • Inhibits tumor growth in male athymic nude mice.
Catalog No. 50-004-39126 Supplier Medchemexpress LLC Supplier No. HY11205510MM/1ML
DIM-C-pPhOH is an antagonist of nuclear receptor 4A1 (NR4A1). It inhibits cancer cell growth, mTOR signaling, and induces apoptosis and cellular stress. It also reduces cell proliferation with IC50 values of 13.6 μM for ACHN cells and 13.0 μM for 786-O cells.
  • NR4A1 antagonist.
  • Inhibits cancer cell growth and mTOR signaling.
  • Induces apoptosis and cellular stress.
  • Reduces cell proliferation in ACHN and 786-O cells.
  • Inhibits NR4A1-regulated expression of survivin, bcl-2, and EGFR.
  • Activates AMPKα and inhibits mTOR.
  • Decreases expression of β1-integrin protein and mRNA.
  • Inhibits migration of MDA-MB-231 and SKBR3 cell lines.
  • Inhibits tumor growth in male athymic nude mice.