Learn More
Medchemexpress LLC HY-112055 5mg Medchemexpress, DIM-C-pPhOH CAS:151358-47-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1120555MG
Encompass_Preferred
Medchemexpress, HY-112055 5mg DIM-C-pPhOH CAS:151358-47-3 DIM-C-pPhOH is a nuclear receptor 4A1 (NR4A1) antagonist. DIM-C-pPhOH inhibits cancer cell growth and mTOR signaling, induce apoptosis and cellular stress. DIM-C-pPhOH reduces cell proliferation with IC50 values of 13.6 μM and 13.0 μM for ACHN cells and 786-O cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9004