Learn More
Medchemexpress LLC DiMal-O-CH2COOH | 1620837-47-9 | 96.8% | 308.24 | C13H12N2O7 | 10 MG
SDP
Catalog No. 5000298746
Encompass_Preferred
This cleavable linker is designed for use in antibody-drug conjugate (ADC) construction and conjugation workflows, offering malemide-based chemistry for attaching payloads to antibodies or targeting moieties. It is provided at research-grade purity and supplied in small quantities suitable for method development and exploratory studies.
  • Cleavable linker for ADC construction and conjugation workflows.
  • Provides maleimide-based chemistry for reliable payload attachment.
  • High purity (96.8%) suitable for research applications.
  • Available in small research quantities; larger amounts available on request.
  • Store at 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-98746 Supplier Medchemexpress LLC Supplier No. HY4922310MG
This cleavable linker is designed for use in antibody-drug conjugate (ADC) construction and conjugation workflows, offering malemide-based chemistry for attaching payloads to antibodies or targeting moieties. It is provided at research-grade purity and supplied in small quantities suitable for method development and exploratory studies.
  • Cleavable linker for ADC construction and conjugation workflows.
  • Provides maleimide-based chemistry for reliable payload attachment.
  • High purity (96.8%) suitable for research applications.
  • Available in small research quantities; larger amounts available on request.
  • Store at 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).