Learn More
Medchemexpress LLC diMal-O-CH2COOH | 1620837-47-9 | 96.8% | C13H12N2O7 | 5 MG
SDP
Catalog No. 5000357772
Encompass_Preferred
diMal-O-CH2COOH is a cleavable ADC linker, intended for research use only.
  • Solid appearance, white to off-white color.
  • Shipping at room temperature in continental US.
  • Store at 4°C, protected from light and under nitrogen.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen.
  • Relevant for cancer and cancer targeted therapy research.
Catalog No. 50-003-57772 Supplier Medchemexpress LLC Supplier No. HY492235MG
diMal-O-CH2COOH is a cleavable ADC linker, intended for research use only.
  • Solid appearance, white to off-white color.
  • Shipping at room temperature in continental US.
  • Store at 4°C, protected from light and under nitrogen.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen.
  • Relevant for cancer and cancer targeted therapy research.